The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010359.2 | NP_034489.1 | 1 - 2 | 75 | 133 |
GenBank mRNA | BC044927.1 | - | 1 - 2 | 59 | 133 |