The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011535.3 | NP_035665.2 | 8 - 8 | 4384 | 117 |
NM_198052.2 | NP_932169.1 | 8 - 8 | 4520 | 117 | |
GenBank mRNA | AK078538.1 | - | 1 - 1 | 1613 | 117 |
AK087128.1 | - | 2 - 2 | 2157 | 117 | |
AK157247.1 | - | 7 - 7 | 4083 | 117 | |
AK172118.1 | - | 7 - 7 | 4089 | 117 | |
BB054411.2 | - | 1 - 1 | 433 | 117 | |
BB728182.1 | - | 1 - 1 | 307 | 117 | |
CJ216213.1 | - | 1 - 1 | 158 | 117 |