The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010430.2 | NP_034560.2 | 1 - 2 | 1255 | 152 |
GenBank mRNA | CJ067923.1 | - | 1 - 2 | 145 | 152 |