Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Chmp4c
- Entrez Gene ID
-
66371
- Gene Name
- charged multivesicular body protein 4C
- Gene Aliases
-
2010012P02Rik, 2210015K02Rik, 2310010I16Rik, Shax3, Snf7-3
- Chromosome Location
-
Chr.3: 10366973 - 10391005 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK008733.1 |
- |
- |
354 |
73 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG971691.1 |
- |
1 |
518 |
|
Chr. 3 - 10393800 - 10394632 [+] on Build GRCm38 |
73 |
GenBankmRNA |
CF615726.1 |
- |
1 |
398 |
|
Chr. 3 - 10393920 - 10394778 [+] on Build GRCm38 |
73 |
GenBankmRNA |
AA915341.1 |
- |
1 |
405 |
|
Chr. 3 - 10393912 - 10394416 [+] on Build GRCm38 |
73 |
GenBankmRNA |
CD773590.1 |
- |
1 |
92 |
|
Chr. 3 - 10394227 - 10394871 [+] on Build GRCm38 |
73 |
GenBankmRNA |
BI855291.1 |
- |
1 |
295 |
|
Chr. 3 - 10394023 - 10394510 [+] on Build GRCm38 |
73 |
GenBankmRNA |
AI122129.1 |
- |
1 |
151 |
|
Chr. 3 - 10394167 - 10394599 [+] on Build GRCm38 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
sorting nexin 16 |
Snx16 |
4930522N22Rik, AI117577, AI747675 |
23.0kb
|
Anti-sense |
Chr. - 10417817 - 10440131 on Build GRCm38 |
View Full Product Details