Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A530030E21Rik
Entrez Gene ID
320731
Gene Name
RIKEN cDNA A530030E21 gene
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK079970.1 - 1 - 1 349 87
AK080099.1 - 1 - 1 347 87


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB637249.1 - 1 345 Chr. 16 - 19243527 - 19244171 [+] on Build GRCm38 87
GenBankmRNA BB636386.1 - 1 349 Chr. 16 - 19243523 - 19244224 [+] on Build GRCm38 87


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream immunoglobulin lambda variable 1 Iglv1 1810027O01Rik, Igl-V1 -158.1kb Anti-sense Chr. - 19085017 - 19085460 on Build GRCm38
Downstream immunoglobulin lambda variable 2 Iglv2 Igl-V2 15.6kb Anti-sense Chr. - 19260403 - 19260844 on Build GRCm38

View Full Product Details