Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY725423.1 - 2 - 3 309 Chr. 1 - 82393521 - 82402743 [-] on Build GRCm38 103
GenBankmRNA AK139597.1 - 313 Chr. - - [-] on Build 103


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream insulin receptor substrate 1 Irs1 G972R, IRS-1 -102.1kb Sense Chr. - 82233105 - 82291439 on Build GRCm38
Downstream collagen, type IV, alpha 4 Col4a4 E130010M05Rik, [a]4(IV) 53.1kb Sense Chr. - 82448419 - 82586849 on Build GRCm38

View Full Product Details