Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cdyl2
- Entrez Gene ID
-
75796
- Gene Name
- chromodomain protein, Y chromosome-like 2
- Gene Aliases
-
1700029M19Rik, 4930453I21Rik, Gm20194
- Chromosome Location
-
Chr.8: 116568724 - 116732991 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_029441.3 |
NP_083717.1 |
7 - 7 |
3890 |
113 |
GenBank mRNA |
AK085145.1 |
- |
1 - 1 |
1047 |
113 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU922735.1 |
- |
1 |
332 |
|
Chr. 8 - 116572656 - 116573252 [-] on Build GRCm38 |
113 |
GenBankmRNA |
CN687276.1 |
- |
1 |
310 |
|
Chr. 8 - 116572682 - 116573230 [-] on Build GRCm38 |
113 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
dynein light chain roadblock-type 2 |
Dynlrb2 |
1700009A04Rik, DNLC2B, Dncl2b |
-55.7kb
|
Anti-sense |
Chr. - 116505015 - 116515915 on Build GRCm38 |
Downstream |
predicted gene, 31367 |
Gm31367 |
|
32.0kb
|
Anti-sense |
Chr. - 116605918 - 116610520 on Build GRCm38 |
View Full Product Details