The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167876.1 | NP_001161348.1 | 25 - 26 | 2109 | 120 |
NM_001167878.1 | NP_001161350.1 | 25 - 26 | 2109 | 120 | |
GenBank mRNA | EU099301.1 | - | 25 - 26 | 2109 | 120 |
EU099302.1 | - | 25 - 26 | 2109 | 120 |