Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skint5
Entrez Gene ID
242627
Gene Name
selection and upkeep of intraepithelial T cells 5
Gene Aliases
OTTMUSG00000008560, Skint-5
Chromosome Location
Chr.4: 113477860 - 113999506 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167876.1 NP_001161348.1 45 - 46 3364 101
NM_001167878.1 NP_001161350.1 45 - 46 3364 101
GenBank mRNA EU099301.1 - 45 - 46 3364 101
EU099302.1 - 45 - 46 3364 101

View Full Product Details