Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd50
Entrez Gene ID
99696
Gene Name
ankyrin repeat domain 50
Gene Aliases
AI662170, E430012K20Rik, mKIAA1223, mKIAA3019
Chromosome Location
Chr.3: 38449261 - 38484861 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167883.1 NP_001161355.1 3 - 4 1670 66
GenBank mRNA AK129484.1 - 3 - 4 1655 66

View Full Product Details