Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vmn2r32
- Entrez Gene ID
-
22311
- Gene Name
- vomeronasal 2, receptor 32
- Gene Aliases
-
V2r5
- Chromosome Location
-
Chr.7: 7463969 - 7479973 on Build GRCm38
- Gene Symbol
- Vmn2r42
- Entrez Gene ID
-
22310
- Gene Name
- vomeronasal 2, receptor 42
- Gene Aliases
-
V2r4
- Chromosome Location
-
Chr.7: 8183265 - 8200320 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009493.2 |
NP_033519.1 |
5 - 6 |
1776 |
86 |
GenBank mRNA |
AF011414.1 |
- |
5 - 6 |
1776 |
86 |
- Gene Symbol
- Vmn2r49
- Entrez Gene ID
-
625605
- Gene Name
- vomeronasal 2, receptor 49
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 9976245 - 9992139 on Build GRCm38
View Full Product Details