Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Defa-rs7
Entrez Gene ID
13226
Gene Name
defensin, alpha, related sequence 7
Gene Aliases
CRS4C-2, CRS4C2, CRS4C2b, CRS4C2c, CRS4C2d, CRS4C3a, CRS4C3a2, CRS4C3b, CRS4C3c, CRS4C3d, CRS4C3e, Defcr-rs7
Chromosome Location
Chr.8: 21191573 - 21192550 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007848.2 NP_031874.2 1 - 2 220 76

Gene Symbol
Defa23
Entrez Gene ID
497114
Gene Name
defensin, alpha, 23
Gene Aliases
Defcr23
Chromosome Location
Chr.8: 21055040 - 21056016 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012307.2 NP_001012307.1 1 - 2 220 76
GenBank mRNA AY746428.1 - 1 - 2 179 76
BC125548.1 - 1 - 2 200 76
BC125550.1 - 1 - 2 200 76

Gene Symbol
Gm15299
Entrez Gene ID
100041811
Gene Name
predicted pseudogene 15299
Gene Aliases
ENSMUSG00000074438, OTTMUSG00000019793
Chromosome Location
Chr.8: 21315546 - 21316387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170955.1 NP_001164426.1 1 - 2 76

View Full Product Details