The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011339.2 | NP_035469.1 | 3 - 4 | 303 | 79 |
GenBank mRNA | AF082859.1 | - | 3 - 4 | 266 | 79 |
AK142955.1 | - | 3 - 4 | 305 | 79 | |
BC061138.1 | - | 3 - 4 | 296 | 79 |