Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Try5
- Entrez Gene ID
-
103964
- Gene Name
- trypsin 5
- Gene Aliases
-
1810049H19Rik, Tc, Tesp4
- Chromosome Location
-
Chr.6: 41311232 - 41314710 on Build GRCm38
- Gene Symbol
- Prss1
- Entrez Gene ID
-
114228
- Gene Name
- protease, serine 1 (trypsin 1)
- Gene Aliases
-
Try-1, Try1, Trygn16
- Chromosome Location
-
Chr.6: 41458930 - 41463786 on Build GRCm38
- Gene Symbol
- Try4
- Entrez Gene ID
-
22074
- Gene Name
- trypsin 4
- Gene Aliases
-
0910001B19Rik, Td
- Chromosome Location
-
Chr.6: 41302272 - 41305533 on Build GRCm38
View Full Product Details