Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BC030870
Entrez Gene ID
407795
Gene Name
cDNA sequence BC030870
Gene Aliases
-
Chromosome Location
Chr.8: 65085613 - 65129537 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033217.1 2 - 3 431 108
GenBank mRNA AK018670.1 - 2 - 3 218 108
AK131860.1 - 2 - 3 456 108
AK136454.1 - 2 - 3 306 108
BC030870.1 - 2 - 3 152 108


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB624953.1 - 2 - 3 308 Chr. 8 - 65086302 - 65129415 [-] on Build GRCm38 108
GenBankmRNA BF578881.1 - 447 Chr. - - [-] on Build 108
GenBankmRNA BQ952327.1 - 2 - 3 302 Chr. 8 - 65086174 - 65129408 [-] on Build GRCm38 108
GenBankmRNA AV063955.2 - 724 Chr. - - [-] on Build 108
GenBankmRNA BB624920.1 - 2 - 3 306 Chr. 8 - 65086302 - 65129412 [-] on Build GRCm38 108
GenBankmRNA AA789670.1 - 2 - 3 364 Chr. 8 - 65086586 - 65129470 [-] on Build GRCm38 108
GenBankmRNA BY720884.1 - 2 - 3 218 Chr. 8 - 65086220 - 65129323 [-] on Build GRCm38 108


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream apelin receptor early endogenous ligand Apela ENSMUSG00000074303, Ela, Ende, Gm10664 -48.3kb Sense Chr. - 65028417 - 65037336 on Build GRCm38
Downstream membrane-associated ring finger (C3HC4) 1 March1 2900024D24Rik, BB085186 509.5kb Anti-sense Chr. - 65617922 - 66471637 on Build GRCm38

View Full Product Details