Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir325
- Entrez Gene ID
-
723929
- Gene Name
- microRNA 325
- Gene Aliases
-
Mirn325, mmu-mir-325
- Chromosome Location
-
Chr.X: 105379082 - 105379179 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029759.1 |
|
1 - 1 |
44 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA 5330434G04 gene |
5330434G04Rik |
|
|
Sense |
Chr. - 105372994 - 105391754 on Build GRCm38 |
Downstream |
cysteine-rich perinuclear theca 2 |
Cypt2 |
Ckt1, Cypt2-ps |
120.6kb
|
Anti-sense |
Chr. - 105499772 - 105500639 on Build GRCm38 |
View Full Product Details