Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR8
Entrez Gene ID
554259
Gene Name
toll like receptor 8
Gene Aliases
-
Chromosome Location
Chr.X: 11482589 - 11498803 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135929.1 NP_001129401.1 1 - 1 3019 76
XM_012106133.3 XP_011961523.1 2 - 2 3094 76
GenBank mRNA AM981306.1 - 1 - 1 3019 76
GU936186.1 - 1 - 1 3019 76
GU936187.1 - 1 - 1 3019 76

View Full Product Details