Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRISP3
Entrez Gene ID
100462691
Gene Name
cysteine-rich secretory protein 3
Gene Aliases
-
Chromosome Location
Chr.20: 24026504 - 24048423 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185121.1 NP_001172050.1 5 - 6 483 78
GenBank mRNA GU985271.1 - 5 - 6 483 78

View Full Product Details