Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SESN2
Entrez Gene ID
101110659
Gene Name
sestrin 2
Gene Aliases
-
Chromosome Location
Chr.2: 254621612 - 254639743 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965423.1 XP_027821224.1 9 - 10 1703 64

View Full Product Details