Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRS1
Entrez Gene ID
101102293
Gene Name
insulin receptor substrate 1
Gene Aliases
-
Chromosome Location
Chr.2: 244237006 - 244301615 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093559.2 XP_014949045.2 1 - 2 3734 71

View Full Product Details