Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMA5
Entrez Gene ID
101104946
Gene Name
proteasome 20S subunit alpha 5
Gene Aliases
-
Chromosome Location
Chr.1: 91227100 - 91247936 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002280.4 XP_004002329.1 6 - 7 571 79

View Full Product Details