Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC38A7
Entrez Gene ID
101105770
Gene Name
solute carrier family 38 member 7
Gene Aliases
-
Chromosome Location
Chr.14: 28030099 - 28043418 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977906.1 XP_027833707.1 5 - 6 927 56
XM_027977907.1 XP_027833708.1 6 - 7 1029 56

View Full Product Details