Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53C
Entrez Gene ID
101111018
Gene Name
family with sequence similarity 53 member C
Gene Aliases
-
Chromosome Location
Chr.5: 52103256 - 52112689 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096059.2 XP_014951545.2 4 - 5 1294 75

View Full Product Details