Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNST
Entrez Gene ID
101115637
Gene Name
consortin, connexin sorting protein
Gene Aliases
-
Chromosome Location
Chr.12: 33380643 - 33470441 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099185.2 XP_014954671.2 5 - 6 922 77

View Full Product Details