Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
101103788
Gene Name
IKAROS family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.2: 229352298 - 229529735 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012145231.3 XP_012000621.1 3 - 4 334 74

View Full Product Details