Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
101116420
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
-
Chromosome Location
Chr.14: 49064224 - 49075006 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015168.3 XP_004015217.2 9 - 10 749 55
XM_027977400.1 XP_027833201.1 9 - 10 743 55

View Full Product Details