Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL3
Entrez Gene ID
101120882
Gene Name
mitochondrial ribosomal protein L3
Gene Aliases
-
Chromosome Location
Chr.1: 281477538 - 281535319 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003343.4 XP_004003392.2 7 - 8 885 64

View Full Product Details