Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR20
Entrez Gene ID
101102936
Gene Name
WD repeat domain 20
Gene Aliases
-
Chromosome Location
Chr.18: 67498884 - 67565300 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015102056.2 XP_014957542.2 2 - 3 495 106

View Full Product Details