Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTERF3
Entrez Gene ID
101108318
Gene Name
mitochondrial transcription termination factor 3
Gene Aliases
MTERFD1
Chromosome Location
Chr.9: 89014222 - 89035286 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015097916.2 XP_014953402.2 4 - 5 988 140
XM_027973202.1 XP_027829003.1 4 - 5 886 140

View Full Product Details