Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMACR
Entrez Gene ID
101113530
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
-
Chromosome Location
Chr.16: 41955452 - 41971270 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017063.4 XP_004017112.2 3 - 4 658 81

View Full Product Details