Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPRD1
Entrez Gene ID
101109600
Gene Name
opioid receptor delta 1
Gene Aliases
-
Chromosome Location
Chr.2: 254154405 - 254193455 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965410.1 XP_027821211.1 2 - 3 662 101

View Full Product Details