Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTN1A1
Entrez Gene ID
101107826
Gene Name
butyrophilin subfamily 1 member A1
Gene Aliases
-
Chromosome Location
Chr.20: 34164583 - 34170958 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019064.4 XP_004019113.2 6 - 7 909 75

View Full Product Details