Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMBR
Entrez Gene ID
101116407
Gene Name
neuromedin B receptor
Gene Aliases
-
Chromosome Location
Chr.8: 73242632 - 73250917 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011382.4 XP_004011431.2 1 - 2 420 87

View Full Product Details