Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM6
Entrez Gene ID
101103179
Gene Name
tripartite motif containing 6
Gene Aliases
-
Chromosome Location
Chr.15: 51467891 - 51486028 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016233.4 XP_004016282.1 2 - 3 574 79

View Full Product Details