Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPT2
Entrez Gene ID
497273
Gene Name
angiopoietin 2
Gene Aliases
ang-2
Chromosome Location
Chr.26: 6014375 - 6070125 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021671.4 XP_004021720.1 5 - 6 1466 70

View Full Product Details