Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SF3B5
Entrez Gene ID
101119646
Gene Name
splicing factor 3b subunit 5
Gene Aliases
-
Chromosome Location
Chr.8: 75141991 - 75142716 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011394.4 XP_004011443.2 1 - 2 407 67

View Full Product Details