Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM38
Entrez Gene ID
101114398
Gene Name
tripartite motif containing 38
Gene Aliases
-
Chromosome Location
Chr.20: 34496327 - 34511242 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012101135.2 XP_011956525.1 3 - 4 751 68
XM_012101137.2 XP_011956527.1 3 - 4 636 68
XM_012101138.2 XP_011956528.1 3 - 4 746 68
XM_012101139.2 XP_011956529.1 4 - 5 671 68
XM_012101140.3 XP_011956530.1 4 - 5 666 68

View Full Product Details