Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101117102
Entrez Gene ID
101117102
Gene Name
zinc finger and SCAN domain-containing protein 4
Gene Aliases
ZSCAN4
Chromosome Location
Chr.14: 70356307 - 70357770 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012094894.3 XP_011950284.2 2 - 3 553 137

View Full Product Details