Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICAM3
Entrez Gene ID
101113326
Gene Name
intercellular adhesion molecule 3
Gene Aliases
-
Chromosome Location
Chr.5: 13193300 - 13201306 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969622.1 XP_027825423.1 6 - 7 1792 90

View Full Product Details