Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLIN4
Entrez Gene ID
101114356
Gene Name
perilipin 4
Gene Aliases
-
Chromosome Location
Chr.5: 18095218 - 18106000 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969845.1 XP_027825646.1 4 - 5 367 76
XM_027969846.1 XP_027825647.1 3 - 4 391 76
XM_027969847.1 XP_027825648.1 4 - 5 367 76

View Full Product Details