Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARVA
Entrez Gene ID
101121625
Gene Name
parvin alpha
Gene Aliases
-
Chromosome Location
Chr.15: 43215786 - 43382376 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015101013.2 XP_014956499.2 4 - 5 470 64

View Full Product Details