Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB19
Entrez Gene ID
101106677
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.4: 112704853 - 112717224 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008108.4 XP_004008157.1 3 - 4 572 105

View Full Product Details