Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPPA
Entrez Gene ID
100009087
Gene Name
natriuretic peptide A
Gene Aliases
CDD
Chromosome Location
Chr.Unknown: 2206083 - 2207462 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082262.1 NP_001075731.1 - 208 75
GenBank mRNA M12046.1 - - 208 75

View Full Product Details