Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMP
Entrez Gene ID
100009142
Gene Name
cathelicidin antimicrobial peptide
Gene Aliases
CAP18
Chromosome Location
Chr.9: 16003796 - 16031921 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082305.1 NP_001075774.1 1 - 2 204 81
XM_008260452.2 XP_008258674.1 4 - 5 818 81
XM_017343635.1 XP_017199124.1 3 - 4 237 81
XM_017343636.1 XP_017199125.1 3 - 4 876 81
GenBank mRNA M73998.1 - 1 - 2 204 81

View Full Product Details