Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1A
Entrez Gene ID
100037708
Gene Name
CD1a molecule
Gene Aliases
-
Chromosome Location
Chr.13: 34832303 - 34835095 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089311.1 NP_001082780.1 3 - 4 611 135
GenBank mRNA AF276977.1 - 3 - 4 611 135

View Full Product Details