Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FSHB
Entrez Gene ID
100008946
Gene Name
follicle stimulating hormone subunit beta
Gene Aliases
FSH
Chromosome Location
Chr.1: 169431050 - 169434819 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082171.2 NP_001075640.1 3 - 3 842 91
GenBank mRNA AY614704.2 - 3 - 3 843 91

View Full Product Details