Gene Transcripts

Gene Symbol
Prl6a1
Entrez Gene ID
24657
Gene Name
prolactin family 6, subfamily a, member 1
Gene Aliases
Plpb, Prlpb
Chromosome Location
Chr. 17: 42705730 - 42710574 on Build Rnor_6.0
UniGene ID
Rn.54426
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022176.2 NP_071512.2 5 968
GenBank mRNA BC158563.1 - 5 968
FQ219971.1 - 5 984
FQ221549.1 - 5 985
FQ230053.1 - 5 986
FQ230138.1 - 2 195
M31155.1 - 5 927

View Full Product Details