Gene Transcripts

Gene Symbol
Pla2g4e
Entrez Gene ID
296091
Gene Name
phospholipase A2, group IVE
Gene Aliases
RGD1310595
Chromosome Location
Chr. 3: 111952396 - 112018409 on Build Rnor_6.0
UniGene ID
Rn.233565
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_230487.7 XP_230487.5 1 743

View Full Product Details