Gene Transcripts

Gene Symbol
Lmod2
Entrez Gene ID
296935
Gene Name
leiomodin 2 (cardiac)
Gene Aliases
Chromosome Location
Chr. 4: 51553439 - 51561135 on Build Rnor_6.0
UniGene ID
Rn.16210
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100964.1 NP_001094434.1 2 945
GenBank mRNA AB331240.1 - 2 821
BC128755.1 - 2 945 7104774

Gene Symbol
LOC100909784
Entrez Gene ID
100909784
Gene Name
leiomodin-2-like
Gene Aliases
Chromosome Location
Chr. 4: 51661538 - 51669306 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749707.3 XP_003749755.1 2 1017

View Full Product Details