Gene Transcripts

Gene Symbol
Fbxo22
Entrez Gene ID
300724
Gene Name
F-box protein 22
Gene Aliases
Chromosome Location
Chr. 8: 59770032 - 59786061 on Build Rnor_6.0
UniGene ID
Rn.4284
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037770.1 NP_001032859.1 5 619
GenBank mRNA BC087061.1 - 5 530
BC107670.1 - 5 619 7937607

View Full Product Details